Gene Transcripts
- Gene Symbol
- ANKRD54
- Entrez Gene ID
- 129138
- Gene Name
- ankyrin repeat domain 54
- Gene Aliases
- LIAR
- Chromosome Location
- Chr. 22: 37830854 - 37848994 on Build GRCh38
- UniGene ID
- Hs.135259
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138797.2 |
NP_620152.1
|
6, 7 |
827 |
|
NR_036556.1 |
|
6, 7 |
822 |
|
XM_006724136.1 |
XP_006724199.1
|
5, 6 |
706 |
|
XM_006724137.1 |
XP_006724200.1
|
5, 6 |
767 |
|
XM_011529877.2 |
XP_011528179.1
|
6, 7 |
1250 |
|
XM_011529878.2 |
XP_011528180.1
|
6, 7 |
1289 |
|
XM_011529879.2 |
XP_011528181.1
|
7, 8 |
677 |
|
XM_011529880.2 |
XP_011528182.1
|
5, 6 |
682 |
|
XM_011529882.2 |
XP_011528184.1
|
6, 7 |
609 |
|
XM_011529883.2 |
XP_011528185.1
|
6, 7 |
840 |
|
XM_011529884.2 |
XP_011528186.1
|
6, 7 |
642 |
|
XM_017028590.1 |
XP_016884079.1
|
6, 7 |
1157 |
|
XM_017028591.1 |
XP_016884080.1
|
6, 7 |
605 |
|
XM_017028593.1 |
XP_016884082.1
|
6, 7 |
1092 |
|
GenBank mRNA |
AK127583.1 |
- |
3, 4 |
1137 |
|
BC014641.1 |
- |
6, 7 |
777 |
2988073 |
BC066909.1 |
- |
6, 7 |
794 |
4795922 |
BX401122.2 |
- |
6, 7 |
825 |
|
CR456471.1 |
- |
6, 7 |
787 |
|
View Full Product Details