Gene Transcripts

Gene Symbol
VWA5B1
Entrez Gene ID
127731
Gene Name
von Willebrand factor A domain containing 5B1
Gene Aliases
Chromosome Location
Chr. 1: 20290907 - 20360442 on Build GRCh38
UniGene ID
Hs.205178
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011540693.1 XP_011538995.1 19 3128
GenBank mRNA AK057346.1 - 8 1249

View Full Product Details