Gene Transcripts

Gene Symbol
BTBD16
Entrez Gene ID
118663
Gene Name
BTB domain containing 16
Gene Aliases
C10orf87
Chromosome Location
Chr. 10: 122270745 - 122338180 on Build GRCh38
UniGene ID
Hs.422466
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318189.1 NP_001305118.1 3, 4 439
NM_144587.3 NP_653188.2 3, 4 436
XM_011539239.2 XP_011537541.1 3, 4 990
XM_011539240.2 XP_011537542.1 3, 4 989
XM_011539241.2 XP_011537543.1 3, 4 989
XM_011539242.2 XP_011537544.1 3, 4 990
XM_017015636.1 XP_016871125.1 2, 3 1519
GenBank mRNA AK058088.1 - 3, 4 426
BC109261.1 - 3, 4 410 40025404
BC109262.1 - 3, 4 410 40025414
DB052430.1 - 3, 4 436

View Full Product Details