Gene Transcripts

Gene Symbol
TTC8
Entrez Gene ID
123016
Gene Name
tetratricopeptide repeat domain 8
Gene Aliases
BBS8, RP51
Chromosome Location
Chr. 14: 88824153 - 88877996 on Build GRCh38
UniGene ID
Hs.303055
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001288781.1 NP_001275710.1 10 1017
NM_001288782.1 NP_001275711.1 8 937
NM_001288783.1 NP_001275712.1 9 909
NM_144596.3 NP_653197.2 10 969
NM_198309.3 NP_938051.1 9 939
NM_198310.3 NP_938052.1 8 849
XM_006720035.1 XP_006720098.1 8 1221
XM_006720037.2 XP_006720100.1 7 1131
XM_011536432.1 XP_011534734.1 9 1299
XM_011536433.1 XP_011534735.1 9 1299
XM_011536434.1 XP_011534736.1 8 1209
XM_011536435.2 XP_011534737.1 8 685
XM_017020977.1 XP_016876466.1 8 1221
XM_017020978.1 XP_016876467.1 7 1131
GenBank mRNA AK093891.1 - 8 787
AK124675.1 - 9 819
AK315772.1 - 8 824
AL833901.1 - 7 673
AY366523.1 - 8 795
AY366524.1 - 7 705
AY373972.1 - 8 795
BC001563.1 - 4 287
BC026351.1 - 9 873 4799330
BC095433.1 - 8 777 30368216
BX161472.1 - 7 777
BX248248.1 - 5 381
KU178849.1 - 9 821
KU178850.1 - 8 743
KU178851.1 - 7 653
KU178852.1 - 3 263

View Full Product Details