Gene Transcripts

Gene Symbol
DHX57
Entrez Gene ID
90957
Gene Name
DEAH-box helicase 57
Gene Aliases
DDX57
Chromosome Location
Chr. 2: 38797729 - 38875883 on Build GRCh38
UniGene ID
Hs.468226
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198963.1 NP_945314.1 4 509
XM_011533154.2 XP_011531456.1 4 513
XM_011533155.2 XP_011531457.1 4 511
XM_011533156.2 XP_011531458.1 4 513
XM_011533157.1 XP_011531459.1 4 513
GenBank mRNA AK057423.1 - 3 309
AK296739.1 - 4 548
AK307514.1 - 4 538
BC039270.1 - 3 316
BC060778.1 - 4 509
BC131534.1 - 4 481 40082400

View Full Product Details