Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145913.3 | NP_666018.3 | 5 | 1032 | |
XM_017018910.1 | XP_016874399.1 | 5 | 1068 | ||
GenBank mRNA | AF536216.1 | - | 5 | 1005 | |
AK075263.1 | - | 5 | 690 | ||
AK313788.1 | - | 5 | 1001 | ||
AY081220.1 | - | 5 | 1006 | ||
BC110492.2 | - | 5 | 667 | 40007567 |