Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_147193.2 | NP_671726.2 | 5 | 1607 | |
XM_017000408.1 | XP_016855897.1 | 6 | 1674 | ||
XM_017000409.1 | XP_016855898.1 | 6 | 1674 | ||
XM_017000410.1 | XP_016855899.1 | 6 | 1788 | ||
XM_017000411.1 | XP_016855900.1 | 6 | 1674 | ||
GenBank mRNA | AK090634.1 | - | 5 | 1606 | |
AK093474.1 | - | 3 | 976 | ||
BC101799.1 | - | 3 | 1113 | 8069305 | |
BC104911.1 | - | 3 | 1113 | 8143914 |