Gene Transcripts

Gene Symbol
CHMP4C
Entrez Gene ID
92421
Gene Name
charged multivesicular body protein 4C
Gene Aliases
SNF7-3, Shax3, VPS32C
Chromosome Location
Chr. 8: 81732453 - 81759515 on Build GRCh38
UniGene ID
Hs.183861
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152284.3 NP_689497.1 5 845
GenBank mRNA AB120734.1 - 5 677
AK000049.1 - 5 807
AK314873.1 - 5 844
AY329086.1 - 5 679
BC014321.1 - 5 849 3828410

View Full Product Details