Gene Transcripts

Gene Symbol
ANKRD13B
Entrez Gene ID
124930
Gene Name
ankyrin repeat domain 13B
Gene Aliases
Chromosome Location
Chr. 17: 29589769 - 29614761 on Build GRCh38
UniGene ID
Hs.662164
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152345.4 NP_689558.4 8, 9 1012
XM_005257920.4 XP_005257977.1 8, 9 945
XM_011524319.2 XP_011522621.1 8, 9 1182
XM_011524320.2 XP_011522622.1 8, 9 944
XM_011524321.2 XP_011522623.1 8, 9 944
XM_017024174.1 XP_016879663.1 8, 9 879
XM_017024175.1 XP_016879664.1 8, 9 881
XM_017024176.1 XP_016879665.1 8, 9 811
GenBank mRNA AK092673.1 - 8, 9 829
AK097716.1 - 7, 8 848
BC032554.1 - 8, 9 1059 5557374
BC062597.1 - 7, 8 862

View Full Product Details