Gene Transcripts
- Gene Symbol
- TMEM237
- Entrez Gene ID
- 65062
- Gene Name
- transmembrane protein 237
- Gene Aliases
- ALS2CR4, JBTS14
- Chromosome Location
- Chr. 2: 201620184 - 201643529 on Build GRCh38
- UniGene ID
- Hs.12319
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001044385.2 |
NP_001037850.1
|
9, 10 |
1006 |
|
NM_152388.3 |
NP_689601.2
|
9, 10 |
1171 |
|
GenBank mRNA |
AB053301.1 |
- |
9, 10 |
1173 |
|
AF212043.1 |
- |
4, 5 |
575 |
|
AK090601.1 |
- |
9, 10 |
864 |
|
AK303954.1 |
- |
9, 10 |
993 |
|
AK304395.1 |
- |
7, 8 |
695 |
|
BC013730.1 |
- |
1, 2 |
89 |
|
BC029611.1 |
- |
9, 10 |
919 |
5304046 |
BX538000.1 |
- |
1, 2 |
66 |
|
View Full Product Details