Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001172771.1 | NP_001166242.1 | 3 | 706 | |
NM_001172772.1 | NP_001166243.1 | 3 | 689 | ||
NM_152456.2 | NP_689669.2 | 3 | 706 | ||
XM_011522902.2 | XP_011521204.1 | 3 | 790 | ||
XM_011522903.2 | XP_011521205.1 | 3 | 572 | ||
GenBank mRNA | AK172744.1 | - | 2 | 221 | |
AK307727.1 | - | 2 | 341 | ||
AK314913.1 | - | 2 | 226 | ||
BC029804.1 | - | 2 | 534 | 5192904 | |
CD248375.1 | - | 3 | 607 | ||
CD515627.1 | - | 3 | 711 | ||
EU599219.1 | - | 2 | 151 | ||
EU636994.1 | - | 2 | 151 |