Gene Transcripts

Gene Symbol
CDCA2
Entrez Gene ID
157313
Gene Name
cell division cycle associated 2
Gene Aliases
PPP1R81, Repo-Man
Chromosome Location
Chr. 8: 25458569 - 25511308 on Build GRCh38
UniGene ID
Hs.33366
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317906.1 NP_001304835.1 14 2542
NM_001317907.1 NP_001304836.1 15 2478
NM_152562.3 NP_689775.2 15 2459
XM_006716294.2 XP_006716357.1 15 2474
XM_011544415.1 XP_011542717.1 16 2546
XM_011544416.2 XP_011542718.1 16 2240
XM_011544417.2 XP_011542719.1 16 2297
XM_011544418.1 XP_011542720.1 16 2510
GenBank mRNA AK092164.1 - 10 1075
AK098670.1 - 2 236
AK292505.1 - 14 2274
AK299278.1 - 15 2184
AL833396.1 - 12 1564
AL833627.1 - 14 2236
BC036214.1 - 12 1587 5267276
BC063651.1 - 2 167
BC085609.1 - 2 222
BG354575.1 - 2 243
BX649008.1 - 11 1450

View Full Product Details