Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152589.1 | NP_689802.1 | 13 | 1581 | |
XM_017018886.1 | XP_016874375.1 | 12 | 1518 | ||
XM_017018887.1 | XP_016874376.1 | 13 | 1824 | ||
XM_017018888.1 | XP_016874377.1 | 11 | 1404 | ||
GenBank mRNA | AK093140.1 | - | 13 | 1581 |