Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001308045.1 | NP_001294974.1 | 2 | 394 | |
NM_152699.4 | NP_689912.2 | 2 | 394 | ||
XM_005269310.2 | XP_005269367.1 | 2 | 394 | ||
XM_005269312.2 | XP_005269369.1 | 2 | 394 | ||
XM_011512544.2 | XP_011510846.1 | 3 | 439 | ||
XM_011512545.2 | XP_011510847.1 | 2 | 394 | ||
XM_017005870.1 | XP_016861359.1 | 3 | 394 | ||
GenBank mRNA | AK302305.1 | - | 2 | 394 | |
AK313764.1 | - | 2 | 367 | ||
BC030705.2 | - | 2 | 343 | 4824344 | |
DC399261.1 | - | 2 | 395 |