Gene Transcripts

Gene Symbol
ANKRD44
Entrez Gene ID
91526
Gene Name
ankyrin repeat domain 44
Gene Aliases
PP6-ARS-B
Chromosome Location
Chr. 2: 196962617 - 197310797 on Build GRCh38
UniGene ID
Hs.432706
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195144.1 NP_001182073.1 4 411
NM_153697.2 NP_710181.2 4 411
XM_005246945.1 XP_005247002.1 4 401
XM_005246946.1 XP_005247003.1 4 401
XM_005246947.2 XP_005247004.1 4 401
XM_005246948.2 XP_005247005.1 4 401
XM_006712838.3 XP_006712901.1 4 401
XM_011512148.2 XP_011510450.1 4 401
XM_017005252.1 XP_016860741.1 4 401
GenBank mRNA AK097086.1 - 4 220
AK124039.1 - 4 322
AK301209.1 - 3 332
BC016985.2 - 4 385 4395011
BC050586.2 - 1 37
BC063622.1 - 4 352

View Full Product Details