Gene Transcripts

Gene Symbol
DCUN1D3
Entrez Gene ID
123879
Gene Name
defective in cullin neddylation 1 domain containing 3
Gene Aliases
44M2.4, SCCRO3
Chromosome Location
Chr. 16: 20854925 - 20900358 on Build GRCh38
UniGene ID
Hs.101007
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173475.3 NP_775746.1 2 565
GenBank mRNA AK123719.1 - 2 496
AY364247.1 - 1 305
BC040442.1 - 2 545 5590225
BX648998.1 - 2 447

View Full Product Details