Gene Transcripts

Gene Symbol
DTWD2
Entrez Gene ID
285605
Gene Name
DTW domain containing 2
Gene Aliases
Chromosome Location
Chr. 5: 118836074 - 118989938 on Build GRCh38
UniGene ID
Hs.655891
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308081.1 NP_001295010.1 2, 3 190
NM_173666.3 NP_775937.1 2, 3 411
XM_011543338.2 XP_011541640.2 2, 3 1744
XM_011543340.2 XP_011541642.1 2, 3 243
GenBank mRNA AK091296.1 - 2, 3 351
BC114508.1 - 2, 3 347 40024023
BC114955.1 - 2, 3 347 40024022
BX648369.1 - 2, 3 190
HY055818.1 - 2, 3 412

View Full Product Details