Gene Transcripts

Gene Symbol
NOP9
Entrez Gene ID
161424
Gene Name
NOP9 nucleolar protein
Gene Aliases
C14orf21
Chromosome Location
Chr. 14: 24299854 - 24309126 on Build GRCh38
UniGene ID
Hs.708040
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286367.1 NP_001273296.1 2 442
NM_174913.2 NP_777573.1 2 442
XM_005267385.1 XP_005267442.1 2 427
XM_011536526.1 XP_011534828.1 2 219
XM_011536527.2 XP_011534829.1 2 427
GenBank mRNA AB095941.1 - 2 434
BC025332.1 - 2 417 4549328
BX248018.1 - 2 404
BX647311.1 - 2 409
HY061279.1 - 2 444

View Full Product Details