Gene Transcripts

Gene Symbol
KLF2
Entrez Gene ID
10365
Gene Name
Kruppel like factor 2
Gene Aliases
LKLF
Chromosome Location
Chr. 19: 16324826 - 16328662 on Build GRCh38
UniGene ID
Hs.685136
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016270.3 NP_057354.1 2 961
GenBank mRNA AF134053.1 - 2 863
AF205849.1 - 2 947
AK225820.1 - 2 960
BC071983.1 - 2 950 5480246

View Full Product Details