Gene Transcripts
- Gene Symbol
- DYNLRB1
- Entrez Gene ID
- 83658
- Gene Name
- dynein light chain roadblock-type 1
- Gene Aliases
- BITH, BLP, DNCL2A, DNLC2A, ROBLD1
- Chromosome Location
- Chr. 20: 34516384 - 34540958 on Build GRCh38
- UniGene ID
- Hs.593920
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001281727.1 |
NP_001268656.1
|
2, 3 |
155 |
|
NM_001281728.1 |
NP_001268657.1
|
3, 4 |
234 |
|
NM_001319157.1 |
NP_001306086.1
|
2, 3 |
159 |
|
NM_014183.3 |
NP_054902.1
|
2, 3 |
159 |
|
NM_177953.2 |
NP_808852.1
|
3, 4 |
238 |
|
GenBank mRNA |
AF132750.1 |
- |
2, 3 |
139 |
|
AF161511.1 |
- |
2, 3 |
130 |
|
AF165516.1 |
- |
2, 3 |
112 |
|
AF178431.1 |
- |
1, 2 |
84 |
|
AK026864.1 |
- |
1, 2 |
95 |
|
AK294214.1 |
- |
2, 3 |
117 |
|
AY026513.1 |
- |
1, 2 |
84 |
|
BC002481.1 |
- |
2, 3 |
137 |
3347555 |
BG110161.1 |
- |
2, 3 |
144 |
|
BM791453.1 |
- |
3, 4 |
220 |
|
BQ639804.1 |
- |
3, 4 |
207 |
|
BU163605.1 |
- |
|
178 |
|
CB149456.1 |
- |
2, 3 |
155 |
|
View Full Product Details