Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172138.1 | NP_742150.1 | 6 | 675 | |
GenBank mRNA | AY129148.1 | - | 6 | 675 | |
DQ126337.1 | - | 6 | 675 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_005258765.4 | XP_005258822.1 | 6 | 1027 |