Gene Transcripts

Gene Symbol
CRYGN
Entrez Gene ID
155051
Gene Name
crystallin gamma N
Gene Aliases
Chromosome Location
Chr. 7: 151428832 - 151448790 on Build GRCh38
UniGene ID
Hs.647104
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144727.2 NP_653328.1 3 639
NR_131779.1 3 991
XM_005249952.3 XP_005250009.1 3 639
XM_011515836.1 XP_011514138.1 3 639
XM_011515837.2 XP_011514139.1 3 1018
GenBank mRNA AF445455.1 - 3 482
BC043605.1 - 3 991
BC100878.2 - 3 477 40003204
BC100880.2 - 3 477 40003208
BC100881.2 - 3 477 40003209
EF491784.1 - 1 64

View Full Product Details