Gene Transcripts

Gene Symbol
NTSR1
Entrez Gene ID
4923
Gene Name
neurotensin receptor 1 (high affinity)
Gene Aliases
NTR
Chromosome Location
Chr. 20: 62708770 - 62762771 on Build GRCh38
UniGene ID
Hs.590869
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002531.2 NP_002522.2 1, 2 1093
XM_011528827.2 XP_011527129.1 1, 2 1160
GenBank mRNA AY429106.1 - 1, 2 722
X70070.1 - 1, 2 1094

View Full Product Details