Gene Transcripts

Gene Symbol
SEC61A1
Entrez Gene ID
29927
Gene Name
Sec61 translocon alpha 1 subunit
Gene Aliases
HSEC61, SEC61, SEC61A
Chromosome Location
Chr. 3: 128052369 - 128071683 on Build GRCh38
UniGene ID
Hs.518236
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013336.3 NP_037468.1 2, 3 259
GenBank mRNA AF077032.1 - 2, 3 165
AF084458.1 - 2, 3 165
AF346602.1 - 2, 3 141
AK027423.1 - 2, 3 259
AK074706.1 - 2, 3 259
AK299118.1 - 2, 3 339
AL832821.1 - 2, 3 192
BC002951.1 - 2, 3 166

View Full Product Details