Gene Transcripts
- Gene Symbol
- CYP3A4
- Entrez Gene ID
- 1576
- Gene Name
- cytochrome P450 family 3 subfamily A member 4
- Gene Aliases
- CP33, CP34, CYP3A, CYP3A3, CYPIIIA3, CYPIIIA4, HLP, NF-25, P450C3, P450PCN1
- Chromosome Location
- Chr. 7: 99756960 - 99784188 on Build GRCh38
- UniGene ID
- Hs.728751
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001202855.2 |
NP_001189784.1
|
2, 3 |
280 |
|
NM_017460.5 |
NP_059488.2
|
2, 3 |
280 |
|
XM_017011780.1 |
XP_016867269.1
|
2, 3 |
278 |
|
GenBank mRNA |
AF182273.1 |
- |
2, 3 |
182 |
|
AK298451.1 |
- |
2, 3 |
273 |
|
AK298462.1 |
- |
2, 3 |
279 |
|
AK312967.1 |
- |
2, 3 |
273 |
|
AY606313.2 |
- |
2, 3 |
173 |
|
BC069418.1 |
- |
2, 3 |
223 |
7262313 |
BC101631.1 |
- |
2, 3 |
269 |
8069137 |
D00003.1 |
- |
2, 3 |
233 |
|
DA639071.1 |
- |
2, 3 |
358 |
|
DQ924960.1 |
- |
2, 3 |
173 |
|
J04449.1 |
- |
2, 3 |
241 |
|
M13785.1 |
- |
2, 3 |
233 |
|
M14096.1 |
- |
2, 3 |
241 |
|
M18907.1 |
- |
2, 3 |
263 |
|
X12387.1 |
- |
2, 3 |
242 |
|
View Full Product Details