Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001297576.1 | NP_001284505.1 | 4 | 626 | |
NM_001297577.1 | NP_001284506.1 | 3 | 424 | ||
NM_003768.4 | NP_003759.1 | 2 | 319 | ||
XM_006711599.2 | XP_006711662.1 | 2 | 223 | ||
GenBank mRNA | AK095879.1 | - | 4 | 531 | |
AK311762.1 | - | 2 | 248 | ||
BC002426.2 | - | 2 | 195 | 3346270 | |
BC022554.1 | - | 2 | 306 | 4820225 | |
BT007252.1 | - | 1 | 113 | ||
CR541733.1 | - | 1 | 113 | ||
DA131259.1 | - | 2 | 303 | ||
DA532423.1 | - | 2 | 303 | ||
DA707281.1 | - | 3 | 351 | ||
HY182028.1 | - | 2 | 322 | ||
X86809.1 | - | 2 | 226 | ||
Y13736.1 | - | 2 | 182 |