Gene Transcripts

Gene Symbol
UGT3A2
Entrez Gene ID
167127
Gene Name
UDP glycosyltransferase family 3 member A2
Gene Aliases
Chromosome Location
Chr. 5: 36035017 - 36071358 on Build GRCh38
UniGene ID
Hs.348941
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168316.1 NP_001161788.1 3 716
NM_174914.3 NP_777574.2 4 818
XM_011513988.2 XP_011512290.1 5 860
XM_017009150.1 XP_016864639.1 4 869
GenBank mRNA AK075383.1 - 4 779
AK300751.1 - 3 677
AY358416.1 - 4 753
BC103924.1 - 4 767 40007344
BC103925.1 - 4 767 40007348

View Full Product Details