Gene Transcripts

Gene Symbol
C8orf37
Entrez Gene ID
157657
Gene Name
chromosome 8 open reading frame 37
Gene Aliases
BBS21, CORD16, RP64, smalltalk
Chromosome Location
Chr. 8: 95244913 - 95270602 on Build GRCh38
UniGene ID
Hs.548157
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177965.3 NP_808880.1 3, 4 348
XM_005250799.3 XP_005250856.2 3, 4 1716
GenBank mRNA AK055162.1 - 3, 4 323
BC036855.2 - 3, 4 313
DA473498.1 - 3, 4 348
DB205618.1 - 3, 4 313
GQ240139.1 - 3, 4 323

View Full Product Details