Gene Transcripts

Gene Symbol
SORBS2
Entrez Gene ID
8470
Gene Name
sorbin and SH3 domain containing 2
Gene Aliases
ARGBP2, PRO0618
Chromosome Location
Chr. 4: 185585444 - 185956716 on Build GRCh38
UniGene ID
Hs.481342
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145670.1 NP_001139142.1 8 1252
NM_001145671.2 NP_001139143.1 7 780
NM_001145672.1 NP_001139144.1 6 577
NM_001145673.1 NP_001139145.1 7 1304
NM_001270771.1 NP_001257700.1 9 1261
NM_003603.6 NP_003594.3 7 1094
NM_021069.4 NP_066547.1 6 1041
XM_005263307.1 XP_005263364.1 9 1898
XM_005263308.1 XP_005263365.1 8 1256
XM_005263310.1 XP_005263367.1 8 1187
XM_005263311.1 XP_005263368.1 8 1256
XM_005263312.1 XP_005263369.1 7 1094
XM_005263313.1 XP_005263370.1 8 1256
XM_006714388.1 XP_006714451.1 8 1187
XM_006714390.1 XP_006714453.1 7 1163
XM_017008738.1 XP_016864227.1 10 2372
XM_017008739.1 XP_016864228.1 11 2675
XM_017008740.1 XP_016864229.1 10 2368
XM_017008741.1 XP_016864230.1 10 2372
XM_017008742.1 XP_016864231.1 9 2322
XM_017008743.1 XP_016864232.1 10 2303
XM_017008744.1 XP_016864233.1 10 2582
XM_017008745.1 XP_016864234.1 9 2522
XM_017008746.1 XP_016864235.1 10 2372
XM_017008747.1 XP_016864236.1 10 2513
XM_017008748.1 XP_016864237.1 10 2372
XM_017008749.1 XP_016864238.1 10 1934
XM_017008750.1 XP_016864239.1 10 2372
XM_017008751.1 XP_016864240.1 10 2372
XM_017008752.1 XP_016864241.1 10 2372
XM_017008753.1 XP_016864242.1 10 2372
XM_017008754.1 XP_016864243.1 10 2372
XM_017008755.1 XP_016864244.1 7 1337
XM_017008756.1 XP_016864245.1 6 1040
XM_017008758.1 XP_016864247.1 7 1206
XM_017008759.1 XP_016864248.1 7 1933
XM_017008760.1 XP_016864249.1 7 1163
XM_017008761.1 XP_016864250.1 10 2372
XM_017008762.1 XP_016864251.1 8 1256
XM_017008763.1 XP_016864252.1 7 1206
XM_017008764.1 XP_016864253.1 8 1187
XM_017008765.1 XP_016864254.1 9 1490
XM_017008766.1 XP_016864255.1 7 1163
XM_017008767.1 XP_016864256.1 8 1466
XM_017008768.1 XP_016864257.1 7 1384
XM_017008769.1 XP_016864258.1 8 1187
XM_017008770.1 XP_016864259.1 7 1163
XM_017008771.1 XP_016864260.1 6 1044
XM_017008772.1 XP_016864261.1 7 1094
XM_017008773.1 XP_016864262.1 8 1397
XM_017008775.1 XP_016864264.1 10 2372
GenBank mRNA AB018320.1 - 6 693
AF049884.1 - 7 1058
AF049885.1 - 6 423
AK056628.1 - 9 879
AK225327.1 - 8 1218
AK225812.1 - 7 1304
AK293400.1 - 6 577
AK295742.1 - 7 612
BC011883.1 - 7 747 3677070

View Full Product Details