Gene Transcripts

Gene Symbol
WIPI2
Entrez Gene ID
26100
Gene Name
WD repeat domain, phosphoinositide interacting 2
Gene Aliases
ATG18B, Atg21, CGI-50, WIPI-2
Chromosome Location
Chr. 7: 5190204 - 5233855 on Build GRCh38
UniGene ID
Hs.122363
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033518.1 NP_001028690.1 8 927
NM_001033519.1 NP_001028691.1 7 873
NM_001033520.1 NP_001028692.1 5 766
NM_001278299.1 NP_001265228.1 8 1101
NM_015610.3 NP_056425.1 8 927
NM_016003.3 NP_057087.2 7 873
XM_006715685.1 XP_006715748.1 6 759
XM_017011952.1 XP_016867441.1 6 759
GenBank mRNA AF151808.1 - 5 765
AK023041.1 - 8 887
AK315468.1 - 8 1047
AL080155.1 - 8 928
AY691425.1 - 7 657
AY691426.1 - 5 697
BC004116.2 - 8 870 3842487
BC007596.1 - 7 840 3346710
BC021012.1 - 7 792
BC021068.1 - 7 792
BC021200.2 - 8 868 3957981
CR533530.1 - 8 711

View Full Product Details