Gene Transcripts

Gene Symbol
NLRP12
Entrez Gene ID
91662
Gene Name
NLR family pyrin domain containing 12
Gene Aliases
CLR19.3, FCAS2, NALP12, PAN6, PYPAF7, RNO, RNO2
Chromosome Location
Chr. 19: 53793584 - 53824403 on Build GRCh38
UniGene ID
Hs.631573
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277126.1 NP_001264055.1 6 2746
NM_001277129.1 NP_001264058.1 6 2743
NM_144687.3 NP_653288.1 6 2743
XM_011527480.1 XP_011525782.1 6 2746
XM_011527482.1 XP_011525784.1 6 2746
XM_017027460.1 XP_016882949.1 6 2746
XM_017027461.1 XP_016882950.1 6 2746
XM_017027462.1 XP_016882951.1 5 2572
XM_017027463.1 XP_016882952.1 7 2519
XM_017027464.1 XP_016882953.1 6 2317
XM_017027465.1 XP_016882954.1 7 2527
XM_017027466.1 XP_016882955.1 7 2524
XM_017027467.1 XP_016882956.1 6 2319
GenBank mRNA AF231021.1 - 4 1671
AK095460.1 - 6 2658
AK290772.1 - 6 2525
AY095146.1 - 6 2690
AY116204.1 - 6 2737
AY116205.1 - 6 2737
AY116206.1 - 6 2737
AY116207.1 - 6 2737
AY154467.1 - 6 2690
BC028069.1 - 6 2633 5212737

View Full Product Details