Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286730.1 | NP_001273659.1 | 6 | 896 | |
NM_080546.4 | NP_536856.2 | 6 | 896 | ||
XM_005251855.4 | XP_005251912.1 | 6 | 901 | ||
XM_006717027.3 | XP_006717090.1 | 6 | 709 | ||
XM_006717028.3 | XP_006717091.1 | 6 | 709 | ||
XM_006717029.2 | XP_006717092.1 | 6 | 709 | ||
XM_017014559.1 | XP_016870048.1 | 6 | 884 | ||
XM_017014560.1 | XP_016870049.1 | 7 | 874 | ||
GenBank mRNA | AJ272365.1 | - | 6 | 849 | |
AJ420812.1 | - | 6 | 682 | ||
AK022549.1 | - | 2 | 249 | ||
AK124926.1 | - | 1 | 876 | ||
BC049203.1 | - | 6 | 649 | 5492167 |