Gene Transcripts

Gene Symbol
TIRAP
Entrez Gene ID
114609
Gene Name
TIR domain containing adaptor protein
Gene Aliases
BACTS1, Mal, MyD88-2, wyatt
Chromosome Location
Chr. 11: 126281268 - 126294933 on Build GRCh38
UniGene ID
Hs.537126
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039661.1 NP_001034750.1 5 1069
NM_001318776.1 NP_001305705.1 4 916
NM_001318777.1 NP_001305706.1 4 916
NM_148910.2 NP_683708.1 5 1085
XM_005271399.3 XP_005271456.2 3 814
XM_011542576.2 XP_011540878.1 3 814
XM_017017161.1 XP_016872650.1 2 2437
XM_017017162.1 XP_016872651.1 5 1106
GenBank mRNA AB446477.1 - 2 633
AF378129.1 - 5 1066
AF406652.1 - 2 633
AF410783.1 - 5 1062
AK124298.1 - 4 916
AK313147.1 - 4 909
AY576785.1 - 2 672
AY576786.1 - 2 672
AY576787.1 - 2 672
BC032474.1 - 5 1038 5216171

View Full Product Details