Gene Transcripts

Gene Symbol
HIGD1B
Entrez Gene ID
51751
Gene Name
HIG1 hypoxia inducible domain family member 1B
Gene Aliases
CLST11240, CLST11240-15
Chromosome Location
Chr. 17: 44844281 - 44850480 on Build GRCh38
UniGene ID
Hs.151787
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271880.1 NP_001258809.1 3 302
NM_016438.3 NP_057522.1 2 386
XM_006721946.2 XP_006722009.1 3 280
XM_006721947.3 XP_006722010.1 3 690
XM_006721948.3 XP_006722011.1 3 384
XM_011524891.2 XP_011523193.1 3 784
XM_017024742.1 XP_016880231.1 3 730
XM_017024743.1 XP_016880232.1 3 822
GenBank mRNA AB038021.1 - 2 213
AB038022.1 - 2 213
BC020667.1 - 2 377 4724613
BG573492.1 - 2 378
BM503772.1 - 2 389

View Full Product Details