Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001281431.1 | NP_001268360.1 | 5 | 786 | |
NM_007196.3 | NP_009127.1 | 6 | 946 | ||
NM_144505.2 | NP_653088.1 | 6 | 1081 | ||
NM_144506.2 | NP_653089.1 | 4 | 523 | ||
NM_144507.2 | NP_653090.1 | 3 | 389 | ||
NR_104008.1 | 5 | 683 | |||
GenBank mRNA | AA320217.1 | - | 2 | 198 | |
AB008390.2 | - | 6 | 821 | ||
AB008927.1 | - | 5 | 923 | ||
AB009849.1 | - | 6 | 801 | ||
AF095742.1 | - | 6 | 946 | ||
AY359036.1 | - | 6 | 932 | ||
BC040887.1 | - | 7 | 1257 | 5742016 | |
BM845575.1 | - | 4 | 489 |