Gene Transcripts
- Gene Symbol
- LOC105379417
- Entrez Gene ID
- 105379417
- Gene Name
- ankyrin repeat domain-containing protein 20A2-like
- Gene Aliases
- Chromosome Location
- Chr. Un|NT_167209: 73517 - 117150 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- ANKRD20A11P
- Entrez Gene ID
- 391267
- Gene Name
- ankyrin repeat domain 20 family member A11, pseudogene
- Gene Aliases
- C21orf81
- Chromosome Location
- Chr. 21: 13943769 - 13980444 on Build GRCh38
- UniGene ID
- Hs.364456
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027270.1 |
|
2 |
776 |
|
GenBank mRNA |
AF426257.1 |
- |
2 |
776 |
|
- Gene Symbol
- ANKRD20A5P
- Entrez Gene ID
- 440482
- Gene Name
- ankyrin repeat domain 20 family member A5, pseudogene
- Gene Aliases
- ANKRD20A5
- Chromosome Location
- Chr. 18: 14179097 - 14227050 on Build GRCh38
- UniGene ID
- Hs.632352
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040113.1 |
|
9 |
1225 |
|
GenBank mRNA |
BP201899.1 |
- |
4 |
551 |
|
- Gene Symbol
- LOC102723382
- Entrez Gene ID
- 102723382
- Gene Name
- ankyrin repeat domain-containing protein 20B-like
- Gene Aliases
- Chromosome Location
- Chr. Un|NT_187499: 21889 - 43425 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- LOC105379426
- Entrez Gene ID
- 105379426
- Gene Name
- ankyrin repeat domain-containing protein 20A2
- Gene Aliases
- Chromosome Location
- Chr. 3|NT_167215: 86 - 16759 on Build GRCh38
- UniGene ID
- Hs.679496
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC016022.1 |
- |
6 |
619 |
|
View Full Product Details