Gene Transcripts

Gene Symbol
MS4A6E
Entrez Gene ID
245802
Gene Name
membrane spanning 4-domains A6E
Gene Aliases
Chromosome Location
Chr. 11: 60334882 - 60340968 on Build GRCh38
UniGene ID
Hs.662211
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139249.2 NP_640342.1 2 274
GenBank mRNA AF333758.1 - 2 325
AF354931.1 - 2 274
AF354932.1 - 2 158
AF354933.1 - 2 214
BC034588.1 - 2 188
BC069322.1 - 2 274 7262144
BC101783.1 - 2 334 8069289

View Full Product Details