Gene Transcripts

Gene Symbol
Alas2
Entrez Gene ID
25748
Gene Name
5'-aminolevulinate synthase 2
Gene Aliases
Chromosome Location
Chr. X: 23167576 - 23187356 on Build Rnor_6.0
UniGene ID
Rn.226279
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013197.1 NP_037329.1 1 96
GenBank mRNA AF186775.1 - 139
BC081857.1 - 117 7133382
D86297.1 - 1 96

View Full Product Details