Gene Transcripts

Gene Symbol
Pitx1
Entrez Gene ID
113983
Gene Name
paired-like homeodomain 1
Gene Aliases
Bft
Chromosome Location
Chr. 17: 8873184 - 8884428 on Build Rnor_6.0
UniGene ID
Rn.74248
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053624.2 NP_446076.1 2 549
XM_006253569.2 XP_006253631.1 3 367
GenBank mRNA AB047298.1 - 2 570
BC061966.1 - 2 549 5621094

View Full Product Details