Gene Transcripts

Gene Symbol
Rbp1
Entrez Gene ID
25056
Gene Name
retinol binding protein 1, cellular
Gene Aliases
Chromosome Location
Chr. 8: 106449321 - 106470842 on Build Rnor_6.0
UniGene ID
Rn.902
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012733.4 NP_036865.1 2 309
GenBank mRNA FQ209770.1 - 2 323
FQ209927.1 - 2 293
FQ210835.1 - 2 324
FQ218090.1 - 2 323
M16459.1 - 2 309
M19257.1 - 2 239

View Full Product Details