Gene Transcripts

Gene Symbol
Dcc
Entrez Gene ID
25311
Gene Name
DCC netrin 1 receptor
Gene Aliases
Chromosome Location
Chr. 18: 66518213 - 67629801 on Build Rnor_6.0
UniGene ID
Rn.10666
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012841.1 NP_036973.1 2 694
XM_008772087.1 XP_008770309.1 1 346
XM_008772088.1 XP_008770310.1 1 346
GenBank mRNA U68725.1 - 2 694

View Full Product Details