Gene Transcripts

Gene Symbol
Mras
Entrez Gene ID
25482
Gene Name
muscle RAS oncogene homolog
Gene Aliases
Chromosome Location
Chr. 8: 107629028 - 107656851 on Build Rnor_6.0
UniGene ID
Rn.10641
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012981.2 NP_037113.2 2, 3 378
GenBank mRNA BC085752.1 - 2, 3 378
D89863.1 - 786

View Full Product Details