Gene Transcripts

Gene Symbol
Scg5
Entrez Gene ID
25719
Gene Name
secretogranin V (7B2 protein)
Gene Aliases
7B2, Sgne1
Chromosome Location
Chr. 3: 105235066 - 105279475 on Build Rnor_6.0
UniGene ID
Rn.6173
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013175.3 NP_037307.1 2 232
XM_006234701.2 XP_006234763.1 2 308
GenBank mRNA BC061560.1 - 2 236
FQ212495.1 - 2 232
FQ213081.1 - 2 258
FQ213100.1 - 2 258
M63901.1 - 2 230

View Full Product Details