Gene Transcripts

Gene Symbol
Cel
Entrez Gene ID
24254
Gene Name
carboxyl ester lipase
Gene Aliases
Bal, Bssl
Chromosome Location
Chr. 3: 7134021 - 7141522 on Build Rnor_6.0
UniGene ID
Rn.91234
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016997.2 NP_058693.2 2, 3 246
GenBank mRNA M15893.1 - 2, 3 246
X16054.1 - 2, 3 242
Z22803.1 - 1, 2 165

View Full Product Details