Gene Transcripts

Gene Symbol
Mcpt1
Entrez Gene ID
29265
Gene Name
mast cell protease 1
Gene Aliases
Vch
Chromosome Location
Chr. 15: 34862249 - 34864803 on Build Rnor_6.0
UniGene ID
Rn.202971
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017145.2 NP_058841.1 2 173
GenBank mRNA AF063851.1 - 2 174
FQ220114.1 - 2 173
FQ220196.1 - 2 173
FQ220424.1 - 2 135
FQ220659.1 - 2 173
FQ220808.1 - 2 206
FQ221313.1 - 2 173
FQ228378.1 - 2 173
FQ229265.1 - 2 173
FQ229320.1 - 2 173
FQ229582.1 - 2 170
FQ229760.1 - 2 173

View Full Product Details