Gene Transcripts

Gene Symbol
Hrct1
Entrez Gene ID
100039781
Gene Name
histidine rich carboxyl terminus 1
Gene Aliases
3830408D24Rik
Chromosome Location
Chr. 4: 43727198 - 43728110 on Build GRCm38
UniGene ID
Mm.18796
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027511.1 NP_081787.1 1 85
GenBank mRNA AK014425.1 - 86

View Full Product Details