Gene Transcripts

Gene Symbol
Gm12942
Entrez Gene ID
100039968
Gene Name
predicted gene 12942
Gene Aliases
OTTMUSG00000009332, Zmym6nb
Chromosome Location
Chr. 4: 127126043 - 127129660 on Build GRCm38
UniGene ID
Mm.392655
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099319.2 NP_001092789.1 3 1077
GenBank mRNA AK034325.1 - 1 3560
AK156456.1 - 3 1059
AK160613.1 - 1 3554

View Full Product Details