Gene Transcripts

Gene Symbol
Igip
Entrez Gene ID
109169
Gene Name
IgA inducing protein
Gene Aliases
6330403M23Rik
Chromosome Location
Chr. 18: 36300101 - 36301478 on Build GRCm38
UniGene ID
Mm.426423
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267796.1 NP_001254725.1 1 1332
GenBank mRNA AK078053.1 - 1 1332

View Full Product Details