Gene Transcripts

Gene Symbol
V1ra8
Entrez Gene ID
113850
Gene Name
vomeronasal 1 receptor, A8
Gene Aliases
Vmn1r-ps33
Chromosome Location
Chr. 6: 90202817 - 90203656 on Build GRCm38
UniGene ID
Mm.377146
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053223.1 NP_444453.1 1 248
GenBank mRNA BC138392.1 - 1 272
BC138394.1 - 1 272 8861414

View Full Product Details