Gene Transcripts

Gene Symbol
Gli2
Entrez Gene ID
14633
Gene Name
GLI-Kruppel family member GLI2
Gene Aliases
AW546128
Chromosome Location
Chr. 1: 118834061 - 119053619 on Build GRCm38
UniGene ID
Mm.273292
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081125.1 NP_001074594.1 4 635
GenBank mRNA AM262743.1 - 3 249
AM262744.1 - 5 466
BC082604.1 - 4 576 30672479
BC085190.1 - 4 551 30694633
X99104.1 - 4 395

View Full Product Details